Learn More
Fisherbrand™ Disposable Aluminum Dishes with Fluted Sides
Useful as evaporating and general utility dishes
Brand:  Fisherbrand™ 08732108
 View more versions of this product
Product Code. 11774059
  • 21.20 € / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Fisherbrand™ Disposable Aluminum Dishes with Fluted Sides are inexpensive enough for single use; useful as evaporating and general utility dishes
Larger sizes are ideal for food processing applications.
  • Made of lightweight aluminum
  • Flat bottoms
  • Fluted or slightly fluted sides
  • Flanged rims
Specifications
Specifications
Flat
Single-use
Round
Useful as evaporating and general utility dishes
109 mm
200 mL
Aluminum
Weighing Dish
77 mm
109 (Inner Top) mm, 77 mm (Inner Base)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Disposable Aluminum Dishes with Fluted Sides > 200mL