Learn More
Fisherbrand™ Electroporation Cuvettes Plus™
Disposable, color-coded cuvets fit most electroporators. Sterilized by gamma irradiation and supplied individually wrapped.
Brand:  Fisherbrand™ 450152
 View more versions of this product
Product Code. 15552423
  • 97.25 € / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Disposable, color-coded cuvets are compatible with most electroporators.
  • Clear, medical grade polycarbonate for high visibility
  • Polished aluminum lowers arcing frequency
  • Round cap allows single-handed removal
  • Sterilized by gamma irradiation and supplied individually wrapped
  • FB102 has imbedded aluminum electrodes
Specifications
Specifications
Sterile
800 μL
Imbedded aluminum electrodes
Electroporator
Electroporation Cuvet
Polycarbonate
4 mm