Learn More
Fisherbrand™ Electroporation Cuvettes Plus™
Disposable, color-coded cuvets fit most electroporators. Sterilized by gamma irradiation and supplied individually wrapped.
97.25 € - 305.00 €
Specifications
Sterility Sterile
Type Electroporation Cuvet
Material Polycarbonate
Includes Imbedded aluminum electrodes
For Use With (Equipment) Electroporator
View More Specs
Products 3
Product Code Brand Capacity (Metric) Width (Metric) Gap Price Quantity & Availability  
Product Code Brand Capacity (Metric) Width (Metric) Gap Price Quantity & Availability  
15532423
View Documents
Fisherbrand™
450150
90 μL 1 mm
305.00 €
Pack of 50
In Stock
to see stock.
Edge
Add to basket
15542423
View Documents
Fisherbrand™
450151
400 μL 2 mm
304.00 €
Pack of 50
In Stock
to see stock.
Edge
Add to basket
15552423
View Documents
Fisherbrand™
450152
800 μL 4 mm
97.25 €
Pack of 50
In Stock
to see stock.
Edge
Add to basket
Description
Description
Disposable, color-coded cuvets are compatible with most electroporators.
  • Clear, medical grade polycarbonate for high visibility
  • Polished aluminum lowers arcing frequency
  • Round cap allows single-handed removal
  • FB102 has imbedded aluminum electrodes
Specifications
Specifications
Sterile
Polycarbonate
Electroporator
Electroporation Cuvet
Imbedded aluminum electrodes