missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Fluorescence Rectangular Cells: Macro Cells With PTFE Lids
Fit standard spectrophotometer cell holders
569.00 € - 745.00 €
Specifications
Type | Macro Fluorescence Cell |
---|---|
Capacity (Metric) | 3000 μL |
Material | Quartz |
Closure Material | PTFE |
Width (Metric) Interior | 10mm |
Includes: PTFE lid or stopper
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
15216043
|
Fisherbrand™
14958128 |
2/Pk., 8 Pk./Cs. |
569.00 €
Pack of 2 |
In Stock
Log in to see stock. ![]() |
||||||
15296053
|
Fisherbrand™
14958130 |
4/Pk., 4 Pk./Cs. |
745.00 €
Pack of 4 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
Description
Fit standard spectrophotometer cell holders
- All cells are 12.5mm wide
- 10mm path length rectangular cells
- Reduce reagent use with micro cells
- Wavelength range for all cells is 200 to 2500nm
- Quartz windows for transmission from far ultraviolet through the infrared
Specifications
Macro Fluorescence Cell | |
Quartz | |
10mm | |
200 to 2500 nm | |
Standard Spectrophotometer Cell Holder | |
Transmission from far ultraviolet through the infrared |
3000 μL | |
PTFE | |
10 mm | |
45 mm | |
12.5 mm | |
Rectangle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Fluorescence Rectangular Cells: Macro Cells With PTFE Lids