missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Heavy-Duty/Utility Funnels
GREENER_CHOICE

Ideal for continuous lab work and heavy-duty jobs

Brand:  Fisherbrand™ 300800-0314

 View more versions of this product

Product Code. 11907944

  • 174.00 € / Pack of 72
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Ideal for continuous lab work and heavy-duty jobs

  • Durable polypropylene construction with external ridges and ribbing to prevent vapor lock
  • Autoclavable to 120°C (248°F)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

8.7cm
Polypropylene
Autoclavable
0.90 in.
100 mL
11 cm
3.44 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.