Learn More
Fisherbrand™ Heavy-Duty/Utility Funnels
Description
Ideal for continuous lab work and heavy-duty jobs
- Durable polypropylene construction with external ridges and ribbing to prevent vapor lock
- Autoclavable to 120°C (248°F)
Specifications
12.5cm | |
Polypropylene | |
Autoclavable |
210 mL | |
14.1 cm |