missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Multiple-Roll Tape Dispenser
A multiple tape dispenser provides a well-organized means of storing a rainbow assortment of color coding tape.
Brand: Fisherbrand™ 15912
36.15 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Description
A multiple tape dispenser provides a well-organized means of storing a rainbow assortment of color coding tape.
- For dispensing multiple rolls with a 1 in. dia. (76.2mm) core (14 yd. [13m] of tape)
- Accommodate 0.50, 0.75, or 3 in. wide (13, 19, or 25mm) tapes for a total dispensing width of 9.50 in. (24cm)
- Dispensers stay put with a rubbery felt-type material to keep them from sliding
Order Info
Order tapes separately.
Specifications
Multiple-Roll Tape Dispenser | |
Tapes with 1 in. Core | |
Metal | |
For dispensing 9.5 W, rolls with 14 yd. tape |