Learn More
Fisherbrand™ Multiple-Roll Tape Dispenser
GREENER_CHOICE
A multiple tape dispenser provides a well-organized means of storing a rainbow assortment of color coding tape.
54.00 €
Specifications
Product Type Multiple-Roll Tape Dispenser
Material Metal
View More Specs
Products 1
Product Code Brand Holds For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Holds For Use With (Equipment) Price Quantity & Availability  
10092594
GREENER_CHOICE
View Documents
Fisherbrand™
15912
Tapes with 1 in. Core For dispensing 9.5 W, rolls with 14 yd. tape
54.00 €
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
A multiple tape dispenser provides a well-organized means of storing a rainbow assortment of color coding tape.
  • For dispensing multiple rolls with a 3 in. dia. (76.2mm) core (14 yd. [13m] of tape)
  • Accommodate 0.50, 0.75, or 3 in. wide (13, 19, or 25mm) tapes for a total dispensing width of 9.50 in. (24cm)
  • Dispensers stay put with a rubbery felt-type material to keep them from sliding
Order Info
Order tapes separately.
Specifications
Specifications
Multiple-Roll Tape Dispenser
Metal