missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polymethylpentene Graduated Cylinders
GREENER_CHOICE

Calibrated To Contain/To Deliver at 20°C

Brand:  Fisherbrand™ 300450-0250

 View more versions of this product

Product Code. 11987884

  • 77.00 € / Pack of 12
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Calibrated To Contain/To Deliver at 20°C

  • Transparent cylinders with large molded-in graduations
  • Easy to read
  • Large rounded bases ribbed for reinforcement and stability
  • Generous pour spout; 10mL size has a conical top for easier filling and pouring
  • Non-wetting
  • Chemical, heat and impact resistant
  • Window-clear

TRUSTED_SUSTAINABILITY
Specifications

Specifications

250 mL
To Deliver, To Contain
Autoclavable
Circular
PMP
2 mL
2mL
Graduated Cylinder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers