missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Microtiter Tubes
GREENER_CHOICE

Ideal for sample dilution and/or mixing prior to transfer into microtiter plates.

Brand:  Fisherbrand™ 02681385

72.69 EUR valid until 2024-12-31
Use promo code "21757" to get your promotional price.


 View more versions of this product

Product Code. 17157885

  • 87.00 € / Pack of 960
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Ideal for sample dilution and/or mixing prior to transfer into microtiter plates

Also suitable for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagents. Choose plain tubes or tubes with frosted writing area.

  • Superior-grade polypropylene tubes and packing racks are autoclavable and can be frozen to -80°C
  • Nonsterile tubes are certified RNase- and DNase-free
  • Sterile tubes are certified RNase-, DNase-, ATP-, DNA- and pyrogen-free
  • Frosted tubes feature writing area that wraps around tube collar
  • Capacity- 1.2mL

Order Info

Packed in bulk, or in convenient 8 x 12 compact tray packs or robotic racks. Compact Tray PacksSnug, smooth- fitting cover protects samples and enables stacking; With paper grid for easy indentification of contentsRobotic RacksSpecifically designed with 96-well footprint for robotic workstations; One-way lid design with alphanumeric grid enables easy sample identification; Condensation rings inside cover minimize cross-contamination; Tubes fit in straight, upright position

Specifications

Specifications

RNase, DNase-free
1.2 mL
Polypropylene
Robotic Pack
No
Autoclavable
Natural
HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagent
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.