Learn More
Fisherbrand™ Polypropylene Microtiter Tubes
Ideal for sample dilution and/or mixing prior to transfer into microtiter plates.
87.00 € - 197.33 €
Specifications
Autoclavable | Autoclavable |
---|---|
Capacity (Metric) | 1.2 mL |
Color | Natural |
Material | Polypropylene |
For Use With (Application) | HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagent |
Product Code | Brand | Certifications/Compliance | Sterility | Packaging | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Certifications/Compliance | Sterility | Packaging | Price | Quantity & Availability | |||
15891757
|
Fisherbrand™
845-FTPS-FIS |
RNase, DNase, ATP, DNA, pyrogen-free | Sterile | Tray Pack | N/A | ||||
17157885
|
Fisherbrand™
02681385 |
RNase, DNase-free | Non-sterile | Robotic Pack |
87.00 €
Pack of 960 |
Estimated Shipment: 27-03-2025
Log in to see stock. |
|||
Description
Ideal for sample dilution and/or mixing prior to transfer into microtiter plates
Also suitable for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagents. Choose plain tubes or tubes with frosted writing area.
- Superior-grade polypropylene tubes and packing racks are autoclavable and can be frozen to -80°C
- Nonsterile tubes are certified RNase- and DNase-free
- Sterile tubes are certified RNase-, DNase-, ATP-, DNA- and pyrogen-free
- Frosted tubes feature writing area that wraps around tube collar
- Capacity- 1.2mL
Order Info
Packed in bulk, or in convenient 8 x 12 compact tray packs or robotic racks. Compact Tray PacksSnug, smooth- fitting cover protects samples and enables stacking; With paper grid for easy indentification of contentsRobotic RacksSpecifically designed with 96-well footprint for robotic workstations; One-way lid design with alphanumeric grid enables easy sample identification; Condensation rings inside cover minimize cross-contamination; Tubes fit in straight, upright position
Specifications
Autoclavable | |
Natural | |
HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagent |
1.2 mL | |
Polypropylene |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.