Learn More
Fisherbrand™ Porcelain Crucible Covers
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles.
Brand:  Fisherbrand™ FB965Y
 View more versions of this product
Product Code. 13489709
Description
Description
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles
  • Glazed inside and out except for the outside bottom surface
  • With perforated handle
  • Strongly recommended that heating and cooling rates not exceed 200°C per hour and maximum use temperature of 1100°C
Specifications
Specifications
With Perforated handle
Porcelain
Use with Fisherbrand high and wide form porcelain crucible
2.75mm
99mm
Crucible Cover
90mm
Glazed
1150°C
76g
8mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Porcelain Crucible Covers > 99 x 8 x 2.75mm; Inner Dia.: 90mm; 76g; Case wt.: 5.25kg