Learn More
Fisherbrand™ Powder Funnels
GREENER_CHOICE
Parallel stems minimize bridging of powder.
Brand:  Fisherbrand™ 300700-0150
21.53 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 12652435
  • 27.00 € / Pack of 4
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Parallel stems minimize bridging of powder
  • Durable polypropylene
  • External ribbing prevents air locks
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
15cm
27mm
13.7 cm
30mm
750 mL
Polypropylene
Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Powder Funnels > Top dia. x H: 150 x 137mm; Capacity: 750mL