missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Reusable Amber Glass Media Bottles
Use these amber Fisherbrand™ Reusable Glass Media Bottles for a variety of general laboratory applications.
50.05 € - 565.00 €
Specifications
Color | Amber |
---|---|
For Use With (Application) | Media and reagent storage, Holding and dispensing fluids, Chemical storage |
Includes | Screw Cap, Pouring Ring |
Material | Borosilicate Glass (3.3) |
Screw Color | Blue |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15817001
|
Fisherbrand™
SPFS14395A-100 |
100 mL |
163.06 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15837001
|
Fisherbrand™
SPFS14395A-250 |
250 mL |
195.05 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15847001
|
Fisherbrand™
SPFS14395A-500 |
500 mL |
244.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15827001
|
Fisherbrand™
SPFS14395A-1000 |
1000 mL |
354.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
17455353
|
Fisherbrand™
SPFS14395A-2000 |
2000 mL |
50.05 €
Each |
In Stock
Log in to see stock. |
|||||
17465353
|
Fisherbrand™
SPFS14395A-5000 |
5000 mL |
202.00 €
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
17445353
|
Fisherbrand™
SPFS14395A-10000 |
10,000 mL |
565.00 €
Each |
Estimated Shipment: 09-10-2024
Log in to see stock. |
|||||
Description
Fisherbrand™ Reusable Amber Glass Media Bottles with cap feature a convenient wide opening, enhanced graduations and a polypropylene pour ring. Feature clear (natural), drip-free, replaceable polypropylene pour ring.
- Permanent white enamel marking spot
- GL 45 screw thread
- Linerless
- Autoclavable (to 140°C)
- Amber borosilicate glass
- Polypropylene cap
Specifications
Amber | |
Screw Cap, Pouring Ring | |
Blue | |
Non-sterile | |
No | |
Autoclavable | |
Yes | |
Polypropylene | |
Screw Cap |
Media and reagent storage, Holding and dispensing fluids, Chemical storage | |
Borosilicate Glass (3.3) | |
Polypropylene | |
Blue | |
Yes | |
Reusable | |
Amber Media Bottle | |
GL 45 |