Learn More
Fisherbrand™ Screw Caps for Disposable Glass Tubes
267.58 EUR valid until 2024-10-31
Use promo code "21661" to get your promotional price.
Description
Phenolic caps have cemented-in rubber liners
- Autoclavable
- Polypropylene cap is linerless
Specifications
15-415 | |
No | |
Autoclavable | |
Black | |
Non-sterile |
Rubber | |
Screw Cap | |
Phenolic | |
Rubber | |
14-959-35A, 14-959-35B, 14-959-35AA |