missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Screw Caps for Disposable Glass Tubes
Phenolic caps have cemented-in rubber liners
218.00 € - 339.53 €
Specifications
Product Type | Screw Cap |
---|---|
Liner Material | Rubber |
Pre-sterilized | No |
Autoclavable | Autoclavable |
Sterility | Non-sterile |
Product Code | Brand | Color | Material | Closure Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Material | Closure Size | Price | Quantity & Availability | |||
11547133
|
Fisherbrand™
FS73800-13415 |
Black | Phenolic | 13-415 |
336.00 €
Pack of 1000 |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||
11567133
|
Fisherbrand™
FS73800-18415 |
Black | Phenolic | 18-415 |
218.00 €
Pack of 1000 |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||
12337279
|
Fisherbrand™
FS73800-15415 |
Black | Phenolic | 15-415 |
339.53 €
Pack of 1000 |
In Stock
Log in to see stock. |
|||
Description
Phenolic caps have cemented-in rubber liners
- Autoclavable
- Polypropylene cap is linerless
Specifications
Screw Cap | |
No | |
Non-sterile |
Rubber | |
Autoclavable |