Learn More
Fisherbrand™ Square Labels
GREENER_CHOICE
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196º C (-311º F). Our cryogenic storage labels can be printed or written on and will not bleed, smudge or smear. Engineered for you to use in your laser printer.
Brand:  Fisherbrand™ 11010222
 View more versions of this product
Product Code. 17837211
  • 58.00 € / 20 sheets
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Labels can be printed or written on and will not bleed, smudge or smear
  • Engineered for use in laser printers
Specifications
Specifications
Orange
Non-sterile
27 x 1.25 in.
Square label sheet
85 Labels/Sheet
0.64 x 3.17 cm