Learn More
Fisherbrand™ Square Labels
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196º C (-311º F). Our cryogenic storage labels can be printed or written on and will not bleed, smudge or smear. Engineered for you to use in your laser printer.
55.20 € - 69.00 €
Product Code | Brand | Color | Product Type | Quantity | Size (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Product Type | Quantity | Size (Metric) | Price | Quantity & Availability | ||
17807201
|
Fisherbrand™
11010209 |
White | Square label sheet | 52/Sheet | 1.91 x 4.29 cm |
55.20 €
20 sheets |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||
17817201
|
Fisherbrand™
11010210 |
Green | Square label sheet | 156/Pk. | 0.64 x 3.81 cm |
55.20 €
20 sheets |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||
17827201
|
Fisherbrand™
11010211 |
Orange | Square label sheet | 156/Pk. | 0.64 x 3.81 cm |
55.20 €
20 sheets |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||
17837201
|
Fisherbrand™
11010212 |
Red | Square label sheet | 156/Pk. | 0.64 x 3.81cm |
55.20 €
20 sheets |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||
17847201
|
Fisherbrand™
11010213 |
Yellow | Square label sheet | 156/Pk. | 0.64 x 3.81cm |
55.20 €
20 sheets |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||
17867201
|
Fisherbrand™
11010215 |
Assorted | Square label sheet | 156/Pk. | 0.64 x 3.81 cm |
69.00 €
25 sheets |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||
17857201
|
Fisherbrand™
11010214 |
White | Square label sheet | 156/Pk. | 0.64 x 3.81cm |
55.20 €
20 sheets |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||
17837211
|
Fisherbrand™
11010222 |
Orange | Square label sheet | 85 Labels/Sheet | 0.64 x 3.17 cm |
55.20 €
20 sheets |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||
17817211
|
Fisherbrand™
11010220 |
Blue | Square label sheet | 85 Labels/Sheet | 0.64 x 3.17 cm |
55.20 €
20 sheets |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||
17827211
|
Fisherbrand™
11010221 |
Green | Square label sheet | 85 Labels/Sheet | 0.64 x 3.17 cm |
55.20 €
20 sheets |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||
17847211
|
Fisherbrand™
11010223 |
Red | Square label sheet | 85 Labels/Sheet | 0.64 x 3.17 cm |
55.20 €
20 sheets |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||
17857211
|
Fisherbrand™
11010224 |
Yellow | Square label sheet | 85 Labels/Sheet | 0.64 x 3.17 cm |
55.20 €
20 sheets |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||
Description
- Labels can be printed or written on and will not bleed, smudge or smear
- Engineered for use in laser printers
Specifications
Non-sterile |