missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sterile Sampling Bags with Flat-Wire Closures

An economical and efficient way to collect, contain and carry samples

Brand:  Fisherbrand™ 14955187

 View more versions of this product

Product Code. 13158167

  • 131.00 € / Pack of 500
Estimated Shipment: 20-06-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Collect, contain and carry samples in an economical and efficient way

  • Sterile to avoid sample contamination
Specifications

Specifications

24 oz.
Clear
3.50 mil
9 in.
5.5 in.
Flat Wire
Polyethylene
9 x 5.5 in. (22.9 x 14cm)
22.9 cm
14 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.