Learn More
Fisherbrand™ Sterile Sampling Bags with Flat-Wire Closures
An economical and efficient way to collect, contain and carry samples
Brand:  Fisherbrand™ 14955186
 View more versions of this product
Product Code. 17903182
  • 95.00 € / Pack of 500
Edge
Explore more special offers
Description
Description
Collect, contain and carry samples in an economical and efficient way
  • Sterile to avoid sample contamination
Specifications
Specifications
18 oz.
Clear
3 mil
9 in.
4.5 in.
Flat Wire
Polyethylene
9 x 4.5 in. (22.9 x 11.4cm)
22.9 cm
11.4 cm