Learn More
Fisherbrand™ Traceable™ 10-Hour Stopwatches
For general-purpose applications
Brand: Fisherbrand™ 90024-02
140.78 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Fisherbrand™ Traceable™ 10-Hour Stopwatches offer single-action timing, time-in/time-out, continuous timing, and cumulative split functions. For general purpose applications.
- Resolution: 0.01 seconds
- Handheld with rugged ABS plastic case
- Net weight: 2.5 oz. (71g)
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Specifications
Digital Stopwatch | |
Audible | |
0.01 sec. | |
ABS Plastic | |
2.5 in. | |
3 in. | |
0.75 in. | |
1.9 cm | |
2.5 oz. | |
4 Year Silver Oxide water resistant, O-ring sealed ABS Plastic case and lanyard | |
ISO 17025, A2LA, NIST |
10 hr. | |
5e-06 | |
LCD | |
2.5 x 3 x 0.75 in. (6.35 x 7.62 x 1.9 cm) | |
6.35 cm | |
7.62 cm | |
0.375 in. | |
0.95 cm | |
0.07 kg | |
Enzyme tests, viscosity measurements, animal studies and other timed lab tests |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.