Learn More
Fisherbrand™ Traceable™ Workstation Digital Barometer
Innovative barometer stands on the bench
Brand:  Fisherbrand™ 4245
Product Code. 11744206
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Includes: Two AAA alkaline batteries.
Description
Description
Measures atmospheric pressure, graphically displays the barometric trend, and reports temperature and time. Ideal for recording ambient changes in the lab, monitoring conditions affecting sample analysis, and assisting in the prediction of weather changes.
  • Graphic barometric trend shows present reading and previous 1-, 3-, 6-, and 12-hour readings in a bar-chart format
  • Forecasting indicators show weather conditions for next 12 hours using symbols for sunny, cloudy, rainy, and partly cloudy
Order Info
Measures 4 x 4 x 2.25 in. (10.2 x 10.2 x 5.7cm). Weight: 4 oz.
Certifications
Traceable to standards provided by NIST™. Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Traceable™ Workstation Digital Barometer > Traceable Digital Workstation Barometer