Learn More
Fisherbrand™ Universal Pipet Pump
Pipet filler with easy one-hand operation provides accurate titration. Color-coded for easy identification.
Brand:  Fisherbrand™ 14955201
 View more versions of this product
Product Code. 15209805
  • 22.55 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Made from high-grade polyethylene
  • Accepts glass and serological pipets
  • Ideal for soap and acid titrations
  • Clear markings make measuring easy
  • 10mL pipet marked in 0.10mL increments
  • 25mL pipet marked in 0.20mL increments
  • Rotating thumbwheel draws or dispensed liquid
  • Side lever allows entire contents to be rapidly dispensed
25mL fits both 10mL and 25mL pipets
Specifications
Specifications
Blue
Polyethylene
Pipet Controller
2 mL