Learn More
Fisherbrand™ Zipper Seal Sample Bags
14.15 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Zipper-locking feature securely seals bags
- Durable, transparent polyethylene
- Ideal for collecting, storing, and protecting laboratory specimens and samples easily and conveniently
- Clear, print-free surface lets user mark and label each bag separately
Not suggested for use with liquids
Specifications
Not Autoclavable | |
Zip Seal | |
LDPE | |
50 | |
8 x 5 in. (20 x 13cm) | |
20 cm | |
13 cm |
1.1 lb. | |
Clear | |
500 | |
4 mil | |
8 in. | |
5 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.