Learn More
Gibco™ Virus Transport Medium
A ready-to-use solution containing Hanks balanced salt solution, heat inactivated FBS, gentamicin sulfate, and amphotericin B.
Brand: Gibco™ A48750DK
This item is not returnable.
View return policy
Description
Kit for transporting samples suspected to contain virus
Gibco Viral Transport Medium is suitable for the transport of samples suspected to contain virus. It is a ready-to-use solution containing Hanks balanced salt solution, heat inactivated FBS, gentamicin sulfate, and amphotericin B.
Specifications
Animal Origin | |
Liquid | |
Standard Serum Supplementation | |
No Glucose, No Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate |
1 X | |
Virus Transport Medium | |
Sterile-filtered | |
Dry Ice |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 9
Product Content Correction
Product Title
Gibco™ Virus Transport Medium > 1 L
Spot an opportunity for improvement?Share a Content Correction