missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BINDER™ Shelf, Reinforced
Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system
Brand: BINDER™ 8012-0288
This item is not returnable.
View return policy
Description
- Reinforced
- Positioned at bottom level
- Maximum load of 45 kg
Specifications
Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system | |
Max load of 67 kg | |
Reinforced Shelf |
BINDER Chambers KB 115 | |
Stainless Steel |