Learn More
Flash Point Standard - Cleveland Open Cup, ASTM D 92 (nominal value: 112 °C), Paragon Scientific™
Specifically designed for use in regular instrument performance checks. Packed uniquely in 3 x 80L single shot bottles assuring the highest level of sample integrity. Typically used in the analysis of petroleum products with flash points above 79°C (175°F) and below 400°C (752°F)
Brand: Paragon Scientific FP-COC-6
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Packaged in 3 X 80mL bottles
- One bottle per test ensures zero contamination / high end volatile loss
- Fully traceable to International Standards
- Supplied with MSDS
- Tamper evident security packaging
- 12 Month shelf life
- Manufactured in the United Kingdom
Specifications
3 x 80 mL |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Nominal values quoted. Certified Values will vary from batch to batch.