Learn More
Thermo Scientific™ Flat PCR Caps, strips of 12
Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.
Brand:  Thermo Scientific™ AB0851
 View more versions of this product
Product Code. 12109840
  • 99.50 € / Pack of 200
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Flat PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights:
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Natural
Polypropylene
25 packs of 8
200 Strip Caps
PCR Plate Cap Strip
96-well Plate
Certified DNA/RNase Free
12
Thermal Cyclers
Thermal Cyclers
Yes