missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Flat Tray in ABS

Flat Tray in ABS can be used in a wide range of temperatures and its dimensions are suitable for refrigerators.

Brand:  Gilac™ G154400

 View more versions of this product

Product Code. 17968631

  • 16.10 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Largest and most customisable range on the market.
  • Can be used in a wide range of temperatures.
  • Dimensions suitable for refrigerators.
  • Can be used as a:
    • defrosting tray thanks to its wide dividing optional grid that separates the ice water from the product being defrosted.
    • preparation container for products that drip.
    • ingredients tray.
    • container for preserving comparison samples.
    • fermenting and decanting container.
  • Frosted bottom, matte finish, for less visibility of scratches.
  • Also available in HDPE, in 5 colors and 5 sizes and in Gilactiv™ version and biobased material.
Specifications

Specifications

8 L
80 mm
HDPE
334 mm
3.1 in.
2.1 gal.
Natural
495 mm
Dispensing Tray
19.5 in.
13.1 in.
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers