missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Flavin adenine dinucleotide disodium salt
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Yellow to Orange |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Quantity | 1 g |
Solubility Information | Solubility in water: 5g/L |
Spectrophotometry | 90% (lambda min. 450nm, pH 7), molar absorptivity 11300 +/- 4%, on dry substance |
Formula Weight | 829.52 |
Physical Form | Powder |
Chemical Name or Material | Flavin adenine dinucleotide disodium salt |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0469131 | Certificate of Analysis | 31/01/2025 | 347210010,347212500 |
A0467878 | Certificate of Analysis | 31/01/2025 | 347210010,347212500 |
A0467880 | Certificate of Analysis | 31/01/2025 | 347210010,347212500 |
A0467879 | Certificate of Analysis | 31/01/2025 | 347210010,347212500 |
A0468913 | Certificate of Analysis | 31/01/2025 | 347210010,347212500 |
Safety and Handling
Product Identifier
- Flavin adenine dinucleotide disodium salt
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals Flavin adenine dinucleotide disodium salt >
Spot an opportunity for improvement?Share a Content Correction