missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Flavin adenine dinucleotide disodium salt Product Code.: 10409301

Thermo Scientific Chemicals Flavin adenine dinucleotide disodium salt

Product Code. 10409301
250 mg, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10409301

Brand: Thermo Scientific Chemicals 347212500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84366-81-4
Molecular Formula C27H31N9Na2O15P2
Molecular Weight (g/mol) 829.52
MDL Number MFCD00151217
InChI Key XLRHXNIVIZZOON-SJFBGTSINA-L
Synonym flavin adenine dinucleotide disodium, unii-67u7uhj04c, flavin adenine dinucleotide disodium salt hydrate, riboflavin 5'-adenosine diphosphate disodium salt, flavin adenine dinucleotide disodium salt, fad-na2, flavine adenine dinucleotide, fad sodium, flavin adenine dinucelotide, fad tnShow MoreShow Less
PubChem CID 131675332
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-4-oxido-2-oxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] phosphate;hydrateShow MoreShow Less
SMILES [Na+].[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)N3C=NC4=C(N)N=CN=C34)C3=NC(=O)NC(=O)C3=NC2=C1Show MoreShow Less

Specifications

Color Yellow to Orange
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: 5g/L
Spectrophotometry 90% (lambda min. 450nm, pH 7), molar absorptivity 11300 +/- 4%, on dry substance
Formula Weight 829.52
Physical Form Powder
Chemical Name or Material Flavin adenine dinucleotide disodium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469131 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number347210010,347212500
Lot NumberA0467878 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number347210010,347212500
Lot NumberA0467880 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number347210010,347212500
Lot NumberA0467879 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number347210010,347212500
Lot NumberA0468913 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number347210010,347212500
Product Identifier
  • Flavin adenine dinucleotide disodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Flavin adenine dinucleotide disodium salt >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.