Learn More
Corning™ Flex Discharge Tubes
Corning™ Flex Discharge Tubes for quick and precise dispensing of acids, bases and solvents
Brand:  Corning™ 6878
 View more versions of this product
Product Code. 16872120
  • 26.55 € / Each
Estimated Shipment: 10-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Coiled
  • Includes protective sleeve
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flexible Discharge Tube
FEP, Polypropylene
Quick and precise dispensing of acids, bases and solvents
Protective Sleeve
Coiled
25 to 100 mL
25 to 100 mL Bottle Top Dispenser
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Flex Discharge Tubes > 25 to 100 mL