missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Description | For Use With (Equipment) | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Size | Price | Quantity & Availability | ||||
12200519
|
Thermo Scientific™
342000000025 |
External Pressure Reducer | ThermoFlex™ 900 - 10000 with P1, P2 or T1 Pumps | 1/2 in. NPT |
477.38 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15681765
|
Thermo Scientific™
342000000027 |
External Pressure Reducer | ThermoFlex™ 7500 - 24000 with P3, P5, T5 and T9 Pumps | 1 in. NPT |
654.26 €
Each |
Estimated Shipment: 27-06-2025
Log in to see stock. |
||||
15560163
|
Thermo Scientific™
611000000106 |
External Flow Control Kit | ThermoFlex™ 900 - 10000 with P1, P2 or T1 Pumps | 1/2 in. NPT |
597.98 €
Each |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||
15510435
|
Thermo Scientific™
342000000029 |
External Flow Control Kit | ThermoFlex™ 7500 - 24000 with P3, P5, T5 and T9 Pumps | 1 in. NPT |
837.17 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Customize the flow and pressure output of recirculating chiller to match the needs of your process. Available in several popular sizes Thermo Scientific™ Flow and pressure control kits for Thermo Scientific chillers can have you up and running without the hassle of finding parts.
- Designed to work with your Thermo Scientific recirculating chiller
- Available in popular sizes that match your recirculating chiller connections and interface with your process
- Take the guess work out of finding parts to match your recirculating chiller connections
- Designed to match the operating range of your recirculating chiller
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Temperature Flow and Pressure Control Accessories