Learn More
Masterflex™ Flow Tubes
Brand:  Masterflex™ 32047-50
 View more versions of this product
Product Code. 15277218
  • 108.00 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Flowtube
2.5S CFH
1/8 in. NPT (F)
150 mm
121°C
±2% (Full Scale)
200 psig
Direct-Reading Flowtube for Air
±0.25%