missing translation for 'onlineSavingsMsg'
Learn More
Flt-3 Inhibitor, Thermo Scientific Chemicals Product Code.: 15426419

Flt-3 Inhibitor, Thermo Scientific Chemicals

Product Code. 15426419
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 15426419

Brand: Thermo Scientific Alfa Aesar J64484.MA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cell-permeable thienylcarboxamide compound that acts as a potent, ATP-competitive, and highly selective Flt-3 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A cell-permeable thienylcarboxamide compound that acts as a potent, ATP-competitive, and highly selective Flt-3 inhibitor
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 301305-73-7
Molecular Formula C18H20N2O4S, C18H20N2O4S
Synonym 2-(3,4-Dimethoxybenzoylamino)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide

Specifications

Quantity 10 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable thienylcarboxamide compound that acts as a potent, ATP-competitive, and highly selective Flt-3 inhibitor
Formula Weight 360.43g/mol
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Powder
InChI Key FSPQCTGGIANIJZ-UHFFFAOYSA-N
SMILES COC1=C(OC)C=C(C=C1)C(=O)NC1=C(C(N)=O)C2=C(CCCC2)S1
IUPAC Name 2-(3,4-dimethoxybenzamido)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
Molecular Weight (g/mol) 360.43
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX16E047 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ64484.MF,J64484.MA
Lot NumberW23H002 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ64484.MF,J64484.MA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Flt-3 Inhibitor, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.