missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Fluo-4 Direct™ Calcium Assay Kit
Fluo-4 Direct™ Calcium Assay Kit
Brand: Molecular Probes™ F10471
446.88 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
This item is not returnable.
View return policy
Description
The Fluo-4 Direct™ Calcium Assay Kit was formulated to provide a homogeneous fluorescent calcium assay that:1. Easily loads into cells
2. Achieves a large assay window and
3. Suppresses background fluorescence generated from the calcium indicator in complete media with little-to-no impact on the specific cellular fluorescence
This advanced formulation allows the assay to be run in a simple addition only format in the presence of serum-containing media. Fluo-4 Direct™ is the third addition to the Fluo-4 family of calcium detection reagents. Fluo-4 AM and Fluo-4 NW both require media removal before assay detection, but Fluo-4 NW adds convenience by including the PowerLoad™ reagent for ease in cell loading. The Fluo-4 Direct™ Calcium Assay Kit is similar to Fluo-4 NW in that it is formulated with PowerLoad™ for ease in cell loading, but unique in that it can be used in the presence of complete culture media and will efficiently suppress background fluorescence without sacrificing the specific cellular fluorescence generated in the assay.

Specifications
Green | |
Fluorescence | |
Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager | |
Fluorescent Dye-Based | |
Powder | |
Molecular Probes™ | |
Room Temperature |
|
|
Calcium Assay | |
Cell-based calcium flux | |
Visible | |
Chemical | |
10 x 10 mL | |
Nucleus, Organelles, Cytoplasm |
Produktvorschläge
Customers who viewed this item also viewed
Viewing 1-5 of 12
Fluo-4 Direct Calcium Assay Kits
Version: 04-03-2009
File size: 195.7 kB
Manual: Fluo4 Direct Calcium Assay Kits (Japanese)
Version: Fluo4 Direct Calcium Assay Kits_J1_3Apr2009.doc
File size: 428.4 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Molecular Probes™ Fluo-4 Direct™ Calcium Assay Kit > 10 x 10mL
For Research Use Only. Not for use in diagnostic procedures.
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur