missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 410710025
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Fluosilicic acid | |
16961-83-4, 7732-18-5 | |
Plastic bottle | |
2.5 kg | |
H2SiF6 | |
MFCD00036289 | |
OHORFAFFMDIQRR-UHFFFAOYSA-P | |
hexafluorosilicon(2-);hydron | |
21863527 | |
19 to 20.6% (total fluorine) |
Colorless | |
1.2700g/mL | |
1.27 | |
F6H2Si | |
Liquid | |
hexafluorosilicic acid, fluorosilicic acid, dihydrogen hexafluorosilicate, silicate 2-, hexafluoro-, dihydrogen, hexafluorosilicon 2-; hydron, dihydrogen hexafluorosilanediuide, silicofluoridd, hexafluorosilic acid, hydrofluorosilic acid, kieselfluorwasserstoffsaure | |
[H+].[H+].F[Si-2](F)(F)(F)(F)F | |
144.091 | |
144.09 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Fluosilicic acid, 25 wt.% aqueous solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction