Learn More
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
33.97 € - 125.63 €
Specifications
Chemical Name or Material | Fluosilicic acid |
---|---|
Color | Colorless |
CAS | 16961-83-4, 7732-18-5 |
Density | 1.2700g/mL |
Packaging | Plastic bottle |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10637021
|
Thermo Scientific Acros
410715000 |
500 g | Plastic bottle |
33.97 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13478548
|
Thermo Scientific Acros
410710025 |
2.5 kg | Plastic bottle |
125.63 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Fluosilicic acid | |
16961-83-4, 7732-18-5 | |
Plastic bottle | |
F6H2Si | |
Liquid | |
hexafluorosilicic acid, fluorosilicic acid, dihydrogen hexafluorosilicate, silicate 2-, hexafluoro-, dihydrogen, hexafluorosilicon 2-; hydron, dihydrogen hexafluorosilanediuide, silicofluoridd, hexafluorosilic acid, hydrofluorosilic acid, kieselfluorwasserstoffsaure | |
[H+].[H+].F[Si-2](F)(F)(F)(F)F | |
144.091 | |
144.09 |
Colorless | |
1.2700g/mL | |
1.27 | |
H2SiF6 | |
MFCD00036289 | |
OHORFAFFMDIQRR-UHFFFAOYSA-P | |
hexafluorosilicon(2-);hydron | |
21863527 | |
19 to 20.6% (total fluorine) |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 241-034-8
missing translation for 'rtecsNumber' : VV8225000
missing translation for 'tsca' : TSCA
- Fluosilicic acid, 25 wt.% aqueous solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.