missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Fmoc-L-Tyrosine (tBu), 10/Pk.
These are Amino Acid Cartridges for the ABI™ 431-433 Peptide Synthesizer.
Brand: Applied Biosystems™ 410650
This item is not returnable.
View return policy
Description
- For Research Use Only
- Not for use in diagnostics procedures

Specifications
Each package contains 10 cartridges. | |
433 A Peptide Synthesizer | |
Peptide Synthesis Reagents |
Cartridge | |
Protein Sequencing & Peptide Synthesis | |
10 Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Applied Biosystems™ Fmoc-L-Tyrosine (tBu), 10/Pk. > 433A Peptide Synthesizer
Spot an opportunity for improvement?Share a Content Correction