missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ White Polyurethane Foam Stoppers

Used as convenient alternative method for closing sterile media flasks and vials instead of cotton plugs.

Brand:  Fisherbrand™ PFS/50X50

 View more versions of this product

Product Code. 11522563

  • 44.00 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Used for microbiology applications
  • During storage of media, it is preferable to cover the stoppers with aluminum foil to avoid dust contamination on removal
  • Shrinkage will occur after continued use following autoclaving, but this is dependent purely on application
  • To eliminate the possibility of obstructing the flow of air into the media container, care should be taken not to crease the stoppers on fitting
Specifications

Specifications

50 mm
50 mm
50 x 50mm
Polyurethane
Microbiology
100/Pk.
White
Plug Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ White Polyurethane Foam Stoppers > Outer Diameter: 50mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.