Learn More
Velp Scientifica™ FOC 200E Connect Cooled Incubator
Refrigerated thermostat of 200 L that guarantees excellent connectivity and high performance in temperature control with the Auto-Tuning System.
Brand:  Velp Scientifica™ F10300542
2189.91 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 16885233
  • 2270.00 € / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Show More
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Velp Scientifica™ FOC 200E Connect Cooled Incubator > FOC 200E Connect Cooled Incubator; Shelf Capacity: 10 kg (22 lb.); Capacity: 44.6 gal. (169 L)