missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Food Specimen Containers with Lids

Food Specimen Containers with Lids are designed especially for storing food specimen samples complying with HACCP catering standards. Markings do not wash off, even in the machine.

Brand:  Gilac™ G160440

 View more versions of this product

Product Code. 17948851

  • 135.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Pack of food specimen containers.
  • Containers especially for storing food specimen samples complying with HACCP catering standards.
  • Markings do not wash off, even in the machine.
Specifications

Specifications

HACCP Food Container
2.1 gal.
Assorted
5.1 in.
19.3 in.
5 Pk.
13.3 in.
HDPE
8 L
491 x 337 x 130 mm
130 mm
491 mm
Rectangular
337 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.