Learn More
Gilac™ Food Specimen Containers with Lids
Food Specimen Containers with Lids are designed especially for storing food specimen samples complying with HACCP catering standards. Markings do not wash off, even in the machine.
Brand:  Gilac™ G160470
 View more versions of this product
Product Code. 17958821
  • 98.10 € / Each
Edge
Explore more special offers
Description
Description
  • Pack of food specimen containers.
  • Containers especially for storing food specimen samples complying with HACCP catering standards.
  • Markings do not wash off, even in the machine.
Specifications
Specifications
HACCP Food Container
0.8 gal.
Assorted
150 mm
350 mm
9.4 in.
HDPE
3 L
5.9 in.
13.8 in.
7 Pk.
240 mm