missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid, LC/MS grade, Thermo Scientific
Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 mL
2 mL
5 mL
50 mL
10 x 1 mL
Specifications
Specifications
Item Name | Formic Acid |
Form | Liquid |
Reagent Type | Ion Pairing Reagent |
Quantity | 2 mL |
Formulation | 99% Formic Acid |
For Use With | LC/MS |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Formic Acid, LC/MS grade, Thermo Scientific >
Spot an opportunity for improvement?Share a Content Correction