Learn More
Brady™ FreezerBondz™ Cryogenic Matte Polyester Labels
Securely mark flat lab surfaces in liquid nitrogen or freezer environments with the durable B-490 polyester label material. Easily print almost anywhere with a Brady™ handheld label printer.
Brand:  Brady™ 170891
 View more versions of this product
Product Code. 17841799
  • 67.95 € / Pack of 80
Explore more special offers
Description
Description
  • Designed for laboratory use on glass and polypropylene vials, centrifuges, test tubes, straws and slides
  • Withstands solvents, smudges and extreme temperatures, including storage in liquid nitrogen
  • Made with a permanent acrylic adhesive for use on flat frozen surfaces or wrapping around tubes and vials with a 1/8 in. overlap
Specifications
Specifications
Autoclavable
Permanent Acrylic
Matte
Polyester
Labels
Laboratory Labeling
80 Labels/Cartridge
0.0026 in.
3.75 in.
3.8 cm
M5-114-490
Black on White
Die-cut
Thermal Transfer
Non-sterile
BMP51, M511
-320°F to +266°F
3.75 x 1.5 in.
1.5 in.
0.5 in.