Learn More
Brady™ FreezerBondz™ Cryogenic Matte Polyester Laboratory Labels for M6/M7 Printers
Durable Polyester markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Brand:  Brady™ 174352
 View more versions of this product
Product Code. 17878164
  • 68.40 € / Pack of 250
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • B-490 labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen when wrapped around itself by at least 1/8 in.
  • Good print smudge resistance, solvent resistance and high/low temperature performance of -196°C to 100°C (-320°F to 212°F)
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Specifications
Specifications
Autoclavable
M6-76-490
None
Matte
Polyester
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
250 Labels
-320°F to +266°F
0.0026 in.
0.6 x 1.625 in.
1.625 in.
15.24 mm
ASTM D 1000, ASTM G 155
Permanent Acrylic
White
Die-cut
Single-side Printable
Labels
Laboratory Labeling
No
Rectangle
-196°C to +130°C
0.065 mm
0.6 in.
41.28 mm