Learn More
Brady™ FreezerBondz™ Cryogenic Matte Polyester Laboratory Labels for M6/M7 Printers
Durable Polyester markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
52.65 € - 130.00 €
Specifications
Autoclavable Autoclavable
Certifications/Compliance ASTM D 1000, ASTM G 155
Adhesive Type Permanent Acrylic
Coating None
Color White
View More Specs
Products 8
Product Code Brand Dimensions (LxW) Height (Metric) Price Quantity & Availability  
Product Code Brand Dimensions (LxW) Height (Metric) Price Quantity & Availability  
17838124
View Documents
Brady™
174179
0.375 x 1.25 in. 9.53 mm
65.55 €
Pack of 500
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
17878164
View Documents
Brady™
174352
0.6 x 1.625 in. 15.24 mm
68.40 €
Pack of 250
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
17851689
View Documents
Brady™
173904
1 x 1.9 in. 25.4 mm
82.30 €
Pack of 250
17868264
View Documents
Brady™
174081
1.65 x 0.5 in. 41.91 mm
112.00 €
Pack of 500
Estimated Shipment: 25-02-2025
to see stock.
Add to basket
17898184
View Documents
Brady™
174094
1.75 x 1 in. 44.45 mm
130.00 €
Pack of 250
Estimated Shipment: 25-02-2025
to see stock.
Add to basket
17848474
View Documents
Brady™
174083
1.8 x 0.6 in. 45.72 mm
114.00 €
Pack of 250
In Stock
to see stock.
Add to basket
17878144
View Documents
Brady™
173708
2.2 x 1.5 in. 55.88 mm
52.65 €
Pack of 100
In Stock
to see stock.
Add to basket
17828454
View Documents
Brady™
173709
3.875 x 1.5 in. 98.43 mm
79.15 €
Pack of 100
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
Description
Description
  • B-490 labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen when wrapped around itself by at least 1/8 in.
  • Good print smudge resistance, solvent resistance and high/low temperature performance of -196°C to 100°C (-320°F to 212°F)
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Specifications
Specifications
Autoclavable
Permanent Acrylic
White
Die-cut
Single-side Printable
Labels
Laboratory Labeling
No
-320°F to +266°F
0.0026 in.
ASTM D 1000, ASTM G 155
None
Matte
Polyester
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
Rectangle
-196°C to +130°C
0.065 mm