Learn More
Brady™ FREEZERBONDZ™ Polyester Labels
For use in laboratory identification applications such as vials, centrifuge tubes, test tubes, straws and slides.
Brand:  Brady™ 142894
 View more versions of this product
Product Code. 15159452
  • 276.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polyester
  • Applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Provides excellent print smudge resistance, solvent resistance and low-temperature performance
Specifications
Specifications
RoHS compliant to 2002/618/EC MCV amendment to RoHS Directive 2011/65/EU
Acrylic, Permanent
Matte
Thermal Transfer
For laboratory identification
1
1500/Pk.
LabelMark, MarkWare
0.013 mm
48.26 mm
FreezerBondz Labels for the BBP33 Printer
White
Polyester
Label
BBP™33
1500/Pk.
48.3 W x 25.4 mm H
-196°C to +130°C
25.40 x 48.26 mm
25.4 mm