Learn More
Brady™ FREEZERBONDZ™ Polyester Labels
For use in laboratory identification applications such as vials, centrifuge tubes, test tubes, straws and slides.
144.00 € - 276.00 €
Specifications
Certifications/Compliance RoHS compliant to 2002/618/EC MCV amendment to RoHS Directive 2011/65/EU
Description FreezerBondz Labels for the BBP33 Printer
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
View More Specs
Products 2
Product Code Brand Size (Metric) Price Quantity & Availability  
Product Code Brand Size (Metric) Price Quantity & Availability  
15180143
View Documents
Brady™
142940
12.7 mm dia.
144.00 €
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
15159452
View Documents
Brady™
142894
48.3 W x 25.4 mm H
276.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Polyester
  • Applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Provides excellent print smudge resistance, solvent resistance and low-temperature performance
Specifications
Specifications
RoHS compliant to 2002/618/EC MCV amendment to RoHS Directive 2011/65/EU
Acrylic, Permanent
Matte
Thermal Transfer
For laboratory identification
LabelMark, MarkWare
0.013 mm
FreezerBondz Labels for the BBP33 Printer
White
Polyester
Label
BBP™33
-196°C to +130°C