Learn More
Brady™ FreezerBondz™ Ultra Thin Cryogenic Polyester Labels
Durable Freezerbondz™ Polyester (B-492) markers are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Brand:  Brady™ 804788
254.11 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 11588834
  • 373.00 € / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Specialized — Aggressive adhesive B-492 FreezerBondz™ labels are ideal for curved, frozen surfaces like vials, centrifuge samples, tubes, and slides.
  • Durable — Labels withstand chemicals, abrasions, storage in liquid nitrogen, and a high/low temperature range of -320°F to 212°F (-196°C to 100°C).
  • Adhesive — Permanent acrylic adhesive for tight bonding onto smooth, curved surfaces, or wrapping around tubes and vials with a 1/8 in. overlap.
  • Certifications — RoHS compliant to 2011/65/EU (Pass 2015/863).
Specifications
Specifications
Not Autoclavable
THT-152-492-2.5-SC
Permanent Acrylic, Permanent
Matte
Polyester
Laboratory Label
Laboratory identification, R6400 series ribbons, thermal transfer printers
27.94 mm
2
Rectangle
-320°F to +266°F
0.081 mm
1 in.
0.37 in.
RoHS compliant to 2011/65/EU
FreezerBondz Thermal Transfer Printer Labels
White
Label
Thermal Transfer
Non-sterile
Thermal transfer printer
Roll of 2500 Labels
2500/Roll
0.375 x 1 in. (L x W)
-160 to 130°C
12.07 mm
25.40 mm
9.53 mm